Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vab-1
Entrez Gene ID
173794
Gene Name
Ephrin receptor 1
Gene Aliases
CELE_M03A1.1
Chromosome Location
Chr.II: 4572000 - 4591386 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062406.6 NP_494807.1 1 - 2 182 90

View Full Product Details