Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acs-11
Entrez Gene ID
173820
Gene Name
fatty Acid CoA Synthetase family
Gene Aliases
CELE_F41C3.3
Chromosome Location
Chr.II: 4742128 - 4745281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062447.9 NP_494848.3 3 - 4 475 77

View Full Product Details