Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clr-1
Entrez Gene ID
3565419
Gene Name
protein-coding
Gene Aliases
CELE_F56D1.4
Chromosome Location
Chr.II: 5466358 - 5473816 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027037.4 NP_001022208.1 15 - 16 3153 141
NM_001027038.3 NP_001022209.1 16 - 17 3239 141
NM_001136345.4 NP_001129817.1 15 - 16 3148 141
NM_001347284.3 NP_001334206.1 16 - 17 3104 141
NR_144560.1 15 - 16 3031 141

View Full Product Details