Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cyp-44A1
- Entrez Gene ID
-
173936
- Gene Name
- Probable cytochrome P450 CYP44
- Gene Aliases
-
CELE_ZK177.5
- Chromosome Location
-
Chr.II: 5504044 - 5505978 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062651.6 |
NP_495052.1 |
6 - 7 |
841 |
132 |
- Gene Symbol
- npp-24
- Entrez Gene ID
-
3564830
- Gene Name
- Protein npp-24
- Gene Aliases
-
CELE_ZK177.4
- Chromosome Location
-
Chr.II: 5503952 - 5509056 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383836.1 |
|
6 - 7 |
933 |
132 |
View Full Product Details