Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-4
Entrez Gene ID
174045
Gene Name
Histone-lysine N-methyltransferase Suv4-20
Gene Aliases
CELE_C32D5.5
Chromosome Location
Chr.II: 6329838 - 6331341 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062871.6 NP_495272.1 2 - 3 253 80

View Full Product Details