Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-16
Entrez Gene ID
174065
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_K06A1.3
Chromosome Location
Chr.II: 6441428 - 6443994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062895.7 NP_495296.2 5 - 6 791 60

View Full Product Details