Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dpy-10
- Entrez Gene ID
-
36805042
- Gene Name
- Cuticle collagen dpy-10
- Gene Aliases
-
CELE_T14B4.7
- Chromosome Location
-
Chr.II: 6710149 - 6712928 on Build WBcel235
- Gene Symbol
- T14B4.19
- Entrez Gene ID
-
174106
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T14B4.19
- Chromosome Location
-
Chr.II: 6710144 - 6712932 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383870.1 |
|
4 - 5 |
560 |
87 |
View Full Product Details