Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fis-1
Entrez Gene ID
185630
Gene Name
Mitochondrial fission 1 protein
Gene Aliases
CELE_F41G3.4
Chromosome Location
Chr.II: 6755136 - 6755965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062980.4 NP_495381.1 1 - 2 96 118

View Full Product Details