Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqv-7
Entrez Gene ID
174145
Gene Name
UDP-sugar transporter sqv-7
Gene Aliases
CELE_C52E12.3
Chromosome Location
Chr.II: 7010811 - 7012690 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063035.6 NP_495436.1 3 - 4 380 108

View Full Product Details