Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ire-1
Entrez Gene ID
174305
Gene Name
protein-coding
Gene Aliases
CELE_C41C4.4
Chromosome Location
Chr.II: 8112243 - 8126876 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267206.3 NP_001254135.1 2 - 3 239 124
GenBank mRNA AF435952.1 - 1 - 2 102 124

View Full Product Details