Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cut-3
Entrez Gene ID
174347
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_F22B5.3
Chromosome Location
Chr.II: 8441025 - 8446834 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063379.7 NP_495780.1 1 - 2 121 69

View Full Product Details