Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
let-19
Entrez Gene ID
174402
Gene Name
Mediator of RNA polymerase II transcription subunit 13
Gene Aliases
CELE_K08F8.6
Chromosome Location
Chr.II: 8770975 - 8782020 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063465.5 NP_495866.2 8 - 9 2159 103
GenBank mRNA AB223006.1 - 8 - 9 2181 103

View Full Product Details