Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-19
Entrez Gene ID
174512
Gene Name
Nuclear hormone receptor family member nhr-19
Gene Aliases
CELE_E02H1.7
Chromosome Location
Chr.II: 9598180 - 9605526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063665.7 NP_496066.2 5 - 6 501 84

View Full Product Details