Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lov-1
Entrez Gene ID
174576
Gene Name
Location of vulva defective 1
Gene Aliases
CELE_ZK945.9
Chromosome Location
Chr.II: 10109100 - 10121825 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063783.4 NP_496184.3 25 - 26 7048 70

View Full Product Details