Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- sinh-1
- Entrez Gene ID
-
190339
- Gene Name
- Stress-activated map kinase-interacting protein 1 homolog
- Gene Aliases
-
CELE_Y57A10A.20
- Chromosome Location
-
Chr.II: 12228305 - 12238242 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_064195.5 |
NP_496596.3 |
2 - 3 |
652 |
113 |
View Full Product Details