Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-51
Entrez Gene ID
183671
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_C50E10.10
Chromosome Location
Chr.II: 12311860 - 12313660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064213.3 NP_496614.3 1 - 2 467 83

View Full Product Details