Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-26
Entrez Gene ID
190349
Gene Name
Serpentine receptor class epsilon-26
Gene Aliases
CELE_Y57A10C.4
Chromosome Location
Chr.II: 12419133 - 12420586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064236.1 NP_496637.1 4 - 5 882 128

View Full Product Details