Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ace-4
Entrez Gene ID
175074
Gene Name
Carboxylic ester hydrolase
Gene Aliases
CELE_Y48B6A.7
Chromosome Location
Chr.II: 14198046 - 14205460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064561.6 NP_496962.1 12 - 13 1426 67
GenBank mRNA AF025379.2 - 12 - 13 1448 67

View Full Product Details