Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acr-20
Entrez Gene ID
187632
Gene Name
protein-coding
Gene Aliases
CELE_R06A4.10
Chromosome Location
Chr.II: 14373831 - 14380897 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027102.6 NP_001022273.1 3 - 4 324 87
NM_001381654.1 NP_001367183.1 2 - 3 192 87
NM_001381655.1 NP_001366936.1 2 - 3 205 87

View Full Product Details