Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spat-1
Entrez Gene ID
175113
Gene Name
protein-coding
Gene Aliases
CELE_F57C2.6
Chromosome Location
Chr.II: 14528752 - 14534174 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267505.2 NP_001254434.1 6 - 7 788 70

View Full Product Details