Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mxl-2
Entrez Gene ID
175184
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_F40G9.11
Chromosome Location
Chr.III: 186946 - 188131 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064772.6 NP_497173.1 2 - 3 283 64

View Full Product Details