Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpy-1
Entrez Gene ID
175342
Gene Name
protein-coding
Gene Aliases
CELE_M01E10.2
Chromosome Location
Chr.III: 2034823 - 2048007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381759.2 NP_001367968.1 1 - 2 153 75
NM_001384037.1 5 - 6 769 75

View Full Product Details