Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msi-1
Entrez Gene ID
175514
Gene Name
MuSashI (fly neural) family
Gene Aliases
CELE_R10E9.1
Chromosome Location
Chr.III: 3964648 - 3976397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001384060.1 6 - 7 899 112
GenBank mRNA AB036738.1 - 6 - 7 899 112

View Full Product Details