Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-11
Entrez Gene ID
191837
Gene Name
Serpentine receptor class gamma-11
Gene Aliases
CELE_T04A8.2
Chromosome Location
Chr.III: 4680013 - 4681562 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065554.2 NP_497955.2 4 - 5 855 76

View Full Product Details