Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-28
Entrez Gene ID
185658
Gene Name
Zinc metalloproteinase nas-28
Gene Aliases
CELE_F42A10.8
Chromosome Location
Chr.III: 6180962 - 6183218 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065941.5 NP_498342.3 6 - 7 829 111

View Full Product Details