Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpt-3
Entrez Gene ID
175925
Gene Name
Probable 26S proteasome regulatory subunit 6B
Gene Aliases
CELE_F23F12.6
Chromosome Location
Chr.III: 6489793 - 6491282 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066028.5 NP_498429.1 2 - 3 841 90

View Full Product Details