Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acbp-4
Entrez Gene ID
186372
Gene Name
ACB domain-containing protein
Gene Aliases
CELE_F56C9.5
Chromosome Location
Chr.III: 7308295 - 7309109 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066208.3 NP_498609.1 2 - 3 480 94

View Full Product Details