Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fmo-3
Entrez Gene ID
176491
Gene Name
protein-coding
Gene Aliases
CELE_Y39A1A.19
Chromosome Location
Chr.III: 10694590 - 10698864 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351929.3 NP_001338838.1 3 - 4 422 75
NM_066955.6 NP_499356.1 3 - 4 513 75
GenBank mRNA AJ582072.1 - 3 - 4 432 75

View Full Product Details