Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- pmk-1
- Entrez Gene ID
-
191743
- Gene Name
- Mitogen-activated protein kinase pmk-1
- Gene Aliases
-
CELE_B0218.3
- Chromosome Location
-
Chr.IV: 8148794 - 8151278 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068964.6 |
NP_501365.1 |
1 - 2 |
162 |
58 |
- Gene Symbol
- faah-2
- Entrez Gene ID
-
177612
- Gene Name
- Amidase domain-containing protein
- Gene Aliases
-
CELE_B0218.2
- Chromosome Location
-
Chr.IV: 8148788 - 8154861 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068965.5 |
NP_501366.1 |
1 - 2 |
168 |
58 |
View Full Product Details