Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hgrs-1
Entrez Gene ID
177617
Gene Name
Hepatocyte Growth factor-Regulated TK Substrate (HRS) family
Gene Aliases
CELE_C07G1.5
Chromosome Location
Chr.IV: 8201374 - 8206048 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068974.8 NP_501375.2 3 - 4 170 91

View Full Product Details