Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-20
Entrez Gene ID
177821
Gene Name
Uncharacterized protein
Gene Aliases
CELE_W08D2.1
Chromosome Location
Chr.IV: 9812359 - 9815526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380420.1 NP_001366963.1 6 - 7 720 91
GenBank mRNA AF103732.1 - 6 - 7 715 91

View Full Product Details