Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-28
Entrez Gene ID
176895
Gene Name
CUTiclin-Like
Gene Aliases
CELE_F41A4.1
Chromosome Location
Chr.IV: 684497 - 688340 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067570.6 NP_499971.2 1 - 2 184 62

View Full Product Details