Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- xpc-1
- Entrez Gene ID
-
177002
- Gene Name
- XPC (Xeroderma Pigmentosum group C) DNA repair gene homolog
- Gene Aliases
-
CELE_Y76B12C.2
- Chromosome Location
-
Chr.IV: 1993089 - 2009027 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067755.6 |
NP_500156.2 |
8 - 9 |
3000 |
64 |
View Full Product Details