Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-10
Entrez Gene ID
177409
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6BL.26
Chromosome Location
Chr.IV: 6446542 - 6457572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306849.3 NP_001293778.1 9 - 10 1119 114
NM_068584.6 NP_500985.4 8 - 9 1024 114

View Full Product Details