Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
csn-6
Entrez Gene ID
178289
Gene Name
protein-coding
Gene Aliases
CELE_Y67H2A.6
Chromosome Location
Chr.IV: 13300824 - 13304145 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268797.2 NP_001255726.1 3 - 4 1005 97

View Full Product Details