Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sqrd-1
Entrez Gene ID
178374
Gene Name
protein-coding
Gene Aliases
CELE_F02H6.5
Chromosome Location
Chr.IV: 14219860 - 14224440 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268869.4 NP_001255798.1 5 - 6 1089 85
NM_001268870.3 NP_001255799.1 5 - 6 953 85
NM_001268871.3 NP_001255800.1 4 - 5 597 85
NM_001307012.3 NP_001293941.1 4 - 5 744 85
NM_001307302.3 NP_001294231.1 2 - 3 198 85
NM_001307303.3 NP_001294232.1 2 - 3 345 85

View Full Product Details