Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-14
Entrez Gene ID
190887
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_Y105C5B.4
Chromosome Location
Chr.IV: 15891654 - 15894233 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070492.7 NP_502893.2 3 - 4 546 140

View Full Product Details