Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vha-8
Entrez Gene ID
177442
Gene Name
Vacuolar H ATPase
Gene Aliases
CELE_C17H12.14
Chromosome Location
Chr.IV: 6790684 - 6792461 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068639.7 NP_501040.1 4 - 5 688 73

View Full Product Details