Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-42
Entrez Gene ID
185168
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F31F4.7
Chromosome Location
Chr.V: 647188 - 649763 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070877.4 NP_503278.2 6 - 7 975 136

View Full Product Details