Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-52
Entrez Gene ID
183493
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C45H4.12
Chromosome Location
Chr.V: 2145485 - 2146144 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071178.2 NP_503579.2 1 - 2 480 90

View Full Product Details