Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-42
Entrez Gene ID
189363
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_Y5H2B.4
Chromosome Location
Chr.V: 2189586 - 2191222 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071195.2 NP_503596.2 3 - 4 546 118

View Full Product Details