Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-59
Entrez Gene ID
178707
Gene Name
protein-coding
Gene Aliases
CELE_T27B7.1
Chromosome Location
Chr.V: 2276013 - 2278265 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316820.3 NP_001303749.1 3 - 4 620 138
NM_071223.8 NP_503624.2 4 - 5 996 138
GenBank mRNA AF273807.1 - 4 - 5 963 138

View Full Product Details