Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-61
Entrez Gene ID
185500
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_F39G3.1
Chromosome Location
Chr.V: 4742422 - 4745373 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071873.5 NP_504274.2 2 - 3 301 77

View Full Product Details