Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-13
Entrez Gene ID
186766
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_H23N18.1
Chromosome Location
Chr.V: 4908846 - 4911399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071916.7 NP_504317.2 5 - 6 1006 142

View Full Product Details