Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-21
Entrez Gene ID
178886
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C02E7.2
Chromosome Location
Chr.V: 4926671 - 4928257 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071924.4 NP_504325.1 2 - 3 306 135

View Full Product Details