Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
K04A8.21
Entrez Gene ID
186973
Gene Name
Saposin B-type domain-containing protein
Gene Aliases
CELE_K04A8.21
Chromosome Location
Chr.V: 6556653 - 6557144 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072264.4 NP_504665.3 2 - 3 201 87

View Full Product Details