Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-46
Entrez Gene ID
183984
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_E02C12.2
Chromosome Location
Chr.V: 9353247 - 9354497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073022.2 NP_505423.1 3 - 4 381 127

View Full Product Details