Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gsnl-1
Entrez Gene ID
179328
Gene Name
Gelsolin-like protein 1
Gene Aliases
CELE_K06A4.3
Chromosome Location
Chr.V: 9497042 - 9501890 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073047.7 NP_505448.1 5 - 6 828 86

View Full Product Details