Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-5
Entrez Gene ID
191715
Gene Name
Na/Ca eXchangers
Gene Aliases
CELE_Y32F6B.2
Chromosome Location
Chr.V: 10486361 - 10490098 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073289.4 NP_505690.2 6 - 7 1154 84

View Full Product Details