Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-6
Entrez Gene ID
185100
Gene Name
Serpentine receptor class epsilon-6
Gene Aliases
CELE_F28H7.9
Chromosome Location
Chr.V: 10760575 - 10762795 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073346.3 NP_505747.2 1 - 2 124 110

View Full Product Details